Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108770.1 | NP_001102240.1 | 5 | 475 | ||
XM_006243332.2 | XP_006243394.1 | 5 | 459 | |||
XM_006243337.2 | XP_006243399.1 | 5 | 498 | |||
XM_006243341.2 | XP_006243403.1 | 5 | 498 | |||
XM_008766315.1 | XP_008764537.1 | 5 | 719 | |||
XM_008766316.1 | XP_008764538.1 | 5 | 572 | |||
XM_008766317.1 | XP_008764539.1 | 5 | 459 | |||
XM_008766318.1 | XP_008764540.1 | 5 | 459 | |||
XM_008766319.1 | XP_008764541.1 | 5 | 498 | |||
XM_008766320.1 | XP_008764542.1 | 5 | 459 |