Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108816.2 | NP_001102286.1 | 2 | 101 | ||
XM_006256870.2 | XP_006256932.1 | 3 | 2351 | |||
XM_006256873.2 | XP_006256935.1 | 3 | 929 | |||
XM_006256875.2 | XP_006256937.1 | 3 | 2471 | |||
XM_006256876.1 | XP_006256938.1 | 3 | 985 | |||
XM_008773175.1 | XP_008771397.1 | 2 | 3649 | |||
XM_008773176.1 | XP_008771398.1 | 3 | 3332 | |||
XM_008773177.1 | XP_008771399.1 | 3 | 2920 | |||
XM_008773178.1 | XP_008771400.1 | 3 | 3388 | |||
XM_008773179.1 | XP_008771401.1 | 3 | 2396 | |||
XM_008773180.1 | XP_008771402.1 | 3 | 2452 | |||
XM_008773181.1 | XP_008771403.1 | 3 | 1453 |