Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100909849 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751721.3 | XP_003751769.1 | 3100 |
Gene Symbol: Fzd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044251.2 | NP_001037716.1 | 1 | 1672 | ||
XM_006254142.2 | XP_006254204.1 | 2 | 3100 | |||
GenBank mRNA | BC100088.1 | - | 1 | 1675 | 7371338 |
Set Membership: |
G-protein coupled receptor |