Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239282.2 | XP_006239344.1 | 6 | 871 | ||
XM_006239283.2 | XP_006239345.1 | 6 | 529 | |||
XM_006239286.2 | XP_006239348.1 | 5 | 372 | |||
XM_008764300.1 | XP_008762522.1 | 7 | 859 | |||
XM_008764301.1 | XP_008762523.1 | 7 | 862 | |||
XM_008764302.1 | XP_008762524.1 | 5 | 359 |