Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001079.1 | NP_001001079.1 | 1 | 63 |
Gene Symbol: LOC100911893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750438.1 | XP_003750486.1 | 1 | 63 |