Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eltd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022294.1 | NP_071630.1 | 6 | 894 | ||
GenBank mRNA | AF192401.1 | - | 6 | 894 | ||
AF192402.1 | - | 6 | 894 |
Set Membership: |
G-protein coupled receptor |