Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031068.1 | NP_112330.1 | 11 | 1174 | ||
GenBank mRNA | AB292042.1 | - | 11 | 1098 | ||
AB292043.1 | - | 11 | 1087 | |||
AF307852.1 | - | 2 | 91 | |||
BC098014.1 | - | 11 | 1174 | 7454624 | ||
FQ212256.1 | - | 11 | 1376 |