Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031121.1 | NP_112383.1 | 1, 2 | 353 | ||
XM_006234477.1 | XP_006234539.1 | 2, 3 | 357 | |||
XM_008761993.1 | XP_008760215.1 | 2, 3 | 339 | |||
XM_008761994.1 | XP_008760216.1 | 2, 3 | 263 | |||
GenBank mRNA | BC083588.1 | - | 1, 2 | 353 | 7132288 |