Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031973.1 | NP_114179.1 | 3, 4 | 378 | ||
GenBank mRNA | AB038232.1 | - | 3, 4 | 375 | ||
AB048711.1 | - | 3, 4 | 378 | |||
BC078783.1 | - | 3, 4 | 351 | 7110490 |