Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032080.1 | NP_114469.1 | 11 | 1367 | ||
XM_006248373.2 | XP_006248435.1 | 12 | 2112 | |||
XM_006248374.2 | XP_006248436.1 | 13 | 2193 | |||
GenBank mRNA | FQ231148.1 | - | 6 | 635 | ||
FQ233111.1 | - | 6 | 635 | |||
X53428.1 | - | 11 | 1342 | |||
X73653.1 | - | 11 | 1367 |