Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053358.1 | NP_445810.1 | 4 | 585 | ||
XM_006238532.2 | XP_006238594.1 | 4 | 620 | |||
XM_006238533.2 | XP_006238595.1 | 4 | 619 | |||
XM_006238534.2 | XP_006238596.1 | 4 | 619 | |||
XM_006238535.2 | XP_006238597.1 | 4 | 617 | |||
XM_006238536.2 | XP_006238598.1 | 4 | 618 | |||
XM_006238537.2 | XP_006238599.1 | 4 | 616 | |||
XM_006238538.2 | XP_006238600.1 | 4 | 616 | |||
XM_008763902.1 | XP_008762124.1 | 4 | 2273 | |||
XM_008763903.1 | XP_008762125.1 | 5 | 1827 | |||
XM_008763904.1 | XP_008762126.1 | 4 | 2270 | |||
XM_008763905.1 | XP_008762127.1 | 4 | 2270 | |||
XM_008763906.1 | XP_008762128.1 | 4 | 2267 | |||
XM_008763907.1 | XP_008762129.1 | 4 | 2269 | |||
XM_008763908.1 | XP_008762130.1 | 4 | 2267 | |||
XM_008763909.1 | XP_008762131.1 | 4 | 2264 | |||
GenBank mRNA | AF121893.1 | - | 4 | 585 | ||
AY724490.1 | - | 4 | 285 | |||
BC107666.1 | - | 4 | 569 | 7936459 |