Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053360.2 | NP_445812.1 | 15, 16 | 1902 | ||
XM_006256924.2 | XP_006256986.2 | 15, 16 | 1844 | |||
XM_006256929.2 | XP_006256991.2 | 13, 14 | 1670 | |||
XM_006256930.2 | XP_006256992.1 | 10, 11 | 1263 | |||
XM_006256931.1 | XP_006256993.1 | 17, 18 | 2195 | |||
XM_008773199.1 | XP_008771421.1 | 4, 5 | 453 | |||
XM_008773200.1 | XP_008771422.1 | 4, 5 | 393 | |||
XM_008773201.1 | XP_008771423.1 | 4, 5 | 349 | |||
GenBank mRNA | AF131867.1 | - | 13, 14 | 1458 | ||
AF230519.1 | - | 15, 16 | 1714 | |||
AF230520.1 | - | 15, 16 | 1568 | |||
AF255885.1 | - | 3, 4 | 620 | |||
AF255886.1 | - | 11, 12 | 1139 | |||
AF255887.1 | - | 10, 11 | 1033 | |||
AF255888.1 | - | 15, 16 | 1820 | |||
BC070877.1 | - | 15, 16 | 1902 | 7096160 |