Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053491.2 | NP_445943.1 | 12 | 1489 | ||
GenBank mRNA | AJ242649.1 | - | 12 | 1492 | ||
AY325159.1 | - | 14 | 1636 | |||
BC091135.1 | - | 12 | 1489 | 7369357 | ||
FQ210878.1 | - | 12 | 1632 | |||
M62832.1 | - | 4 | 426 |