Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053606.2 | NP_446058.1 | 5 | 662 | ||
GenBank mRNA | AB010960.1 | - | 5 | 823 | ||
BC086586.1 | - | 5 | 662 | 7309721 | ||
FQ220321.1 | - | 5 | 668 | |||
FQ220929.1 | - | 5 | 665 |