Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039691.2 | NP_001034780.1 | 2 | 240 | ||
NM_057105.3 | NP_476446.2 | 1 | 202 | |||
GenBank mRNA | AF359279.1 | - | 1 | 202 | ||
AF461737.1 | - | 1 | 210 | |||
AY435133.1 | - | 1 | 117 | |||
BC107931.1 | - | 1 | 213 | 7126208 | ||
D83796.1 | - | 1 | 117 | |||
J02612.1 | - | 2 | 241 | |||
J05132.1 | - | 2 | 379 | |||
S70355.1 | - | 3 | 180 |