Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ust5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134380.1 | NP_599207.1 | 2 | 643 | ||
XM_008760213.1 | XP_008758435.1 | 2 | 1256 | |||
GenBank mRNA | AJ132858.1 | - | 2 | 643 | ||
BC081730.1 | - | 2 | 962 | |||
BC098905.1 | - | 2 | 619 | 7325342 |