Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019126.2 | NP_061999.2 | 4, 5 | 1335 | ||
GenBank mRNA | U09815.1 | - | 1335 |