Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012893.1 | NP_037025.1 | 2 | 163 | ||
XM_006236728.2 | XP_006236790.1 | 3 | 284 | |||
GenBank mRNA | BC087689.1 | - | 3 | 229 | 7310913 | |
FQ220056.1 | - | 3 | 228 | |||
FQ220823.1 | - | 3 | 229 | |||
FQ221094.1 | - | 3 | 228 | |||
FQ221892.1 | - | 3 | 228 | |||
FQ229904.1 | - | 3 | 229 | |||
M22323.1 | - | 2 | 163 |