Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147165.1 | NP_671694.1 | 5 | 567 | ||
GenBank mRNA | M76733.1 | - | 5 | 567 |