Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105808.1 | NP_001099278.1 | 5, 6 | 2834 | ||
XM_006246866.2 | XP_006246928.1 | 3676 |