Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106041.1 | NP_001099511.1 | 4 | 714 | ||
XM_006252041.1 | XP_006252103.1 | 4 | 626 |