Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106175.1 | NP_001099645.1 | 6 | 945 | ||
XM_006255448.2 | XP_006255510.1 | 5 | 693 |