Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019182.2 | NP_062055.1 | 8 | 729 | ||
XM_008770271.1 | XP_008768493.1 | 8 | 761 | |||
XM_008770272.1 | XP_008768494.1 | 8 | 716 | |||
XM_008770273.1 | XP_008768495.1 | 8 | 849 | |||
GenBank mRNA | AF022081.1 | - | 8 | 647 | ||
AY050655.1 | - | 8 | 741 | |||
BC062024.1 | - | 8 | 729 | 5623495 | ||
FQ211855.1 | - | 7 | 611 |