Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014810.1 | NP_001014810.1 | 4, 5 | 514 | ||
NM_198734.2 | NP_942029.2 | 4, 5 | 524 | |||
XM_008759128.1 | XP_008757350.1 | 4, 5 | 543 | |||
XM_008759129.1 | XP_008757351.1 | 4, 5 | 523 | |||
GenBank mRNA | BC058153.1 | - | 4, 5 | 517 | 6887903 | |
CB576820.1 | - | 4, 5 | 467 | |||
CB751111.1 | - | 1, 2 | 120 |
Gene Symbol: LOC103689986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228658.2 | XP_006228720.1 | 4, 5 | 543 | ||
XM_006228660.2 | XP_006228722.1 | 4, 5 | 523 | |||
XM_008759221.1 | XP_008757443.1 | 4, 5 | 553 | |||
XM_008759222.1 | XP_008757444.1 | 4, 5 | 572 |