Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170573.1 | NP_001164044.1 | 2 | 305 | ||
GenBank mRNA | FM034155.1 | - | 2 | 309 | ||
FQ229851.1 | - | 2 | 240 |
Gene Symbol: LOC100910130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749779.3 | XP_003749827.1 | 2 | 289 |