Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005765.1 | NP_001005765.1 | 4, 5 | 295 | ||
XM_006233076.1 | XP_006233138.1 | 2, 3 | 136 | |||
GenBank mRNA | BC083813.1 | - | 4, 5 | 295 | 7110254 | |
D88312.1 | - | 2, 3 | 133 | |||
FQ233812.1 | - | 3, 4 | 201 |