Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddrgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106512.1 | NP_001099982.1 | 8, 9 | 1210 | ||
GenBank mRNA | FQ214225.1 | - | 8, 9 | 795 | ||
FQ231424.1 | - | 8, 9 | 834 |