Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239406.2 | XP_006239468.1 | 876 | |||
XM_006239407.2 | XP_006239469.1 | 974 | ||||
XM_006239408.2 | XP_006239470.1 | 1000 | ||||
XM_006239409.2 | XP_006239471.1 | 903 |
Gene Symbol: Miip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017450.1 | NP_001017450.1 | 5 | 801 | ||
XM_006239379.2 | XP_006239441.1 | 5 | 876 | |||
XM_006239380.2 | XP_006239442.1 | 5 | 974 | |||
XM_006239381.2 | XP_006239443.1 | 5 | 903 | |||
GenBank mRNA | BC079047.1 | - | 5 | 801 | 7115212 | |
FJ618907.1 | - | 5 | 1005 | |||
FQ227320.1 | - | 5 | 759 |