Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107345.1 | NP_001100815.1 | 3 | 772 | ||
XM_008771570.1 | XP_008769792.1 | 3 | 509 |