Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107510.1 | NP_001100980.1 | 8, 9 | 884 | ||
XM_006229051.2 | XP_006229113.1 | 10, 11 | 1232 | |||
XM_006229052.2 | XP_006229114.1 | 11, 12 | 1153 | |||
XM_006229053.2 | XP_006229115.1 | 10, 11 | 1268 | |||
XM_006229054.2 | XP_006229116.1 | 10, 11 | 1304 | |||
XM_008759366.1 | XP_008757588.1 | 9, 10 | 1045 | |||
XM_008759367.1 | XP_008757589.1 | 10, 11 | 1268 | |||
XM_008759368.1 | XP_008757590.1 | 10, 11 | 1265 | |||
XM_008759369.1 | XP_008757591.1 | 9, 10 | 2097 | |||
GenBank mRNA | BC168932.1 | - | 10, 11 | 1309 |