Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108195.1 | NP_001101665.1 | 2, 3 | 1394 | ||
XM_006244106.2 | XP_006244168.1 | 2, 3 | 1459 |