Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100732.1 | NP_001094202.1 | 2, 3 | 457 | ||
GenBank mRNA | BC082050.1 | - | 390 | |||
BC098710.1 | - | 2, 3 | 459 | |||
BC158874.1 | - | 2, 3 | 457 | 7933460 |