Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108861.1 | NP_001102331.1 | 3 | 214 | ||
XM_006251037.2 | XP_006251099.1 | 3 | 211 | |||
XM_006251038.2 | XP_006251100.1 | 3 | 208 | |||
XM_006251039.2 | XP_006251101.1 | 4 | 278 |