Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002279.1 | NP_001002279.1 | 2 | 325 | ||
GenBank mRNA | AY606065.1 | - | 2 | 325 | ||
BC100057.1 | - | 2 | 324 | 7131133 |