Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022528.2 | NP_071973.1 | 7, 8 | 870 | ||
GenBank mRNA | AJ277827.1 | - | 7, 8 | 871 |