Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031005.3 | NP_112267.1 | 19 | 2541 | ||
GenBank mRNA | AF115386.1 | - | 19 | 2541 | ||
AY437436.1 | - | 19 | 2332 |