Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138831.1 | NP_620186.1 | 4 | 1230 | ||
XM_008772974.1 | XP_008771196.1 | 4 | 1269 | |||
GenBank mRNA | AB047324.1 | - | 4 | 1230 |