Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255269.2 | XP_006255331.1 | 31 | 3251 | ||
XM_006255270.2 | XP_006255332.1 | 31 | 3251 | |||
XM_008772410.1 | XP_008770632.1 | 31 | 3963 | |||
GenBank mRNA | FQ217825.1 | - | 9 | 936 | ||
FQ234320.1 | - | 6 | 899 |