Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorbs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005762.1 | NP_001005762.1 | 19 | 1793 | ||
XM_006252241.2 | XP_006252303.1 | 20 | 3477 | |||
XM_006252242.1 | XP_006252304.1 | 19 | 1884 | |||
XM_006252244.2 | XP_006252306.1 | 18 | 2112 | |||
XM_006252246.1 | XP_006252308.1 | 19 | 1829 | |||
XM_006252248.1 | XP_006252310.1 | 7 | 677 | |||
XM_008770819.1 | XP_008769041.1 | 17 | 3586 | |||
GenBank mRNA | BC083666.1 | - | 19 | 1793 | 7135547 |