Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019180.2 | NP_062053.2 | 5 | 786 | ||
GenBank mRNA | D38455.1 | - | 6 | 796 | ||
U67909.1 | - | 5 | 787 |