Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759265.1 | XP_008757487.1 | 3 | 180 |
Gene Symbol: Polr2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106244.1 | NP_001099714.1 | 3 | 531 |