Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047092.1 | NP_001040557.1 | 1 | 177 | ||
XM_006248930.2 | XP_006248992.1 | 4 | 713 | |||
XM_006248931.2 | XP_006248993.1 | 3 | 600 | |||
XM_006248932.2 | XP_006248994.1 | 2 | 1010 | |||
XM_006248933.2 | XP_006248995.1 | 2 | 927 | |||
XM_008768970.1 | XP_008767192.1 | 2 | 715 | |||
GenBank mRNA | AJ879913.1 | - | 1 | 177 | ||
BC160815.1 | - | 1 | 196 | 7365526 |