Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam228a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014074.1 | NP_001014096.1 | 2 | 339 | ||
XM_006239858.2 | XP_006239920.1 | 2 | 631 | |||
GenBank mRNA | BC083643.1 | - | 2 | 339 | 7134934 |