Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126283.1 | NP_001119755.1 | 2 | 309 | ||
XM_006242525.2 | XP_006242587.1 | 2 | 347 | |||
GenBank mRNA | BC161982.1 | - | 2 | 309 | 9030206 |