Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053345.1 | NP_445797.1 | 2, 3 | 188 | ||
XM_006243380.2 | XP_006243442.1 | 3, 4 | 338 | |||
GenBank mRNA | AF000944.1 | - | 2, 3 | 188 | ||
BC168727.1 | - | 4, 5 | 372 | 6888477 | ||
FQ209546.1 | - | 343 | ||||
FQ212213.1 | - | 3, 4 | 323 | |||
FQ221947.1 | - | 3, 4 | 329 | |||
FQ228685.1 | - | 3, 4 | 343 |