Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008721.1 | NP_001008721.1 | 185 | |||
GenBank mRNA | BC087738.1 | - | 185 | 7310934 | ||
DQ480746.1 | - | 1174 | ||||
FQ213511.1 | - | 206 | ||||
FQ219880.1 | - | 130 | ||||
FQ231568.1 | - | 206 |
Gene Symbol: LOC100912375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750007.3 | XP_003750055.1 | 2 | 205 |