Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170558.1 | NP_001164029.1 | 1 | 757 | ||
NM_001170559.1 | NP_001164030.1 | 1 | 757 | |||
NM_001170560.1 | NP_001164031.1 | 1 | 757 | |||
NM_053663.1 | NP_446115.1 | 2 | 855 | |||
GenBank mRNA | AF072892.1 | - | 757 |