Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130823.3 | NP_570836.1 | 2 | 292 | ||
XM_006245898.1 | XP_006245960.1 | 3 | 793 | |||
GenBank mRNA | BC063154.1 | - | 2 | 296 | 6918203 | |
D10874.1 | - | 2 | 233 | |||
FQ211954.1 | - | 2 | 272 | |||
FQ212145.1 | - | 2 | 297 | |||
FQ212559.1 | - | 2 | 276 | |||
FQ212733.1 | - | 2 | 276 | |||
FQ213105.1 | - | 2 | 276 | |||
FQ213455.1 | - | 2 | 276 | |||
FQ214388.1 | - | 2 | 276 | |||
FQ214503.1 | - | 2 | 297 | |||
FQ216440.1 | - | 2 | 276 | |||
FQ220211.1 | - | 2 | 276 | |||
FQ220713.1 | - | 2 | 297 | |||
FQ220906.1 | - | 2 | 245 | |||
FQ221569.1 | - | 2 | 287 | |||
FQ229281.1 | - | 2 | 277 | |||
FQ229453.1 | - | 2 | 272 | |||
FQ229871.1 | - | 2 | 277 | |||
FQ234225.1 | - | 2 | 297 |