Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133570.5 | NP_598254.2 | 3 | 513 | ||
GenBank mRNA | AW524119.1 | - | 1 | 367 | ||
M31176.1 | - | 3 | 917 |