Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138530.2 | NP_612539.2 | 110 | |||
XM_006256372.2 | XP_006256434.1 | 3 | 425 | |||
GenBank mRNA | AY083160.1 | - | 122 | |||
BC078735.1 | - | 110 | 7126368 | |||
FQ209640.1 | - | 145 |